Thermo Scientific Chemicals

2,5-Dimethylthiophene, 98+%, Thermo Scientific Chemicals

Catalog number: A10869.14
25 g, Each
Thermo Scientific Chemicals

2,5-Dimethylthiophene, 98+%, Thermo Scientific Chemicals

Catalog number: A10869.14
25 g, Each
Quantity
Catalog number: A10869.14
also known as A10869-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
638-02-8
IUPAC Name
2,5-dimethylthiophene
Molecular Formula
C6H8S
InChI Key
GWQOOADXMVQEFT-UHFFFAOYSA-N
SMILES
CC1=CC=C(C)S1
Form
Liquid
Appearance (Color)
Clear colorless to yellow to orange
Refractive Index
1.5105-1.5145 @ 20°C
Identification (FTIR)
Conforms
Assay (GC)
≥98.0%

Description

2,5-Dimethylthiophene is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dimethylthiophene is used as an intermediate in organic synthesis.

Solubility
Insoluble in water. Soluble in alcohol, ether and benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text