Thermo Scientific Chemicals

2,3,5-Triphenyl-2H-tetrazolium chloride, 98%, Thermo Scientific Chemicals

Catalog number: A10870.09
10 g, Each
Thermo Scientific Chemicals

2,3,5-Triphenyl-2H-tetrazolium chloride, 98%, Thermo Scientific Chemicals

Catalog number: A10870.09
10 g, Each
Quantity
Catalog number: A10870.09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
298-96-4
IUPAC Name
triphenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Molecular Formula
C19H15ClN4
InChI Key
PKDBCJSWQUOKDO-UHFFFAOYSA-M
SMILES
[Cl-].C1=CC=C(C=C1)N1N=C(N=[N+]1C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White to pale yellow or cream
Form
Crystals or crystalline powder or powder
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
≤1.0%
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%

Description

2,3,5-Triphenyl-2H-tetrazolium chloride is used as a redox indicator. It is also used in biochemical experiments, especially to indicate cellular respiration. It is employed in autopsy pathology to assist identification of post-mortem myocardial infarctions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,5-Triphenyl-2H-tetrazolium chloride is used as a redox indicator. It is also used in biochemical experiments, especially to indicate cellular respiration. It is employed in autopsy pathology to assist identification of post-mortem myocardial infarctions.

Solubility
Soluble in water, ethanol and acetone. Insoluble in ether.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text