Thermo Scientific™

n-Propyl 3,4,5-trihydroxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: A10877.06
5 g, Each
Thermo Scientific™

n-Propyl 3,4,5-trihydroxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: A10877.06
5 g, Each
Quantity
Catalog number: A10877.06
also known as A10877-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
121-79-9
IUPAC Name
propyl 3,4,5-trihydroxybenzoate
Molecular Formula
C10H12O5
InChI Key
ZTHYODDOHIVTJV-UHFFFAOYSA-N
SMILES
CCCOC(=O)C1=CC(O)=C(O)C(O)=C1
Appearance (Color)
White to pale cream or pale brown
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥97.5% to ≤102.5% (dry weight basis)
Identification (FTIR)
Conforms
Loss on Drying
≤1% (110°C, 4hr)

Description

n-Propyl 3,4,5-trihydroxybenzoate is an anti-fade reagent in fluorescence microscopy to reduce photobleaching of fluorescent probes such as rhodamine and fluorescein. Also used as an antioxidant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Propyl 3,4,5-trihydroxybenzoate is an anti-fade reagent in fluorescence microscopy to reduce photobleaching of fluorescent probes such as rhodamine and fluorescein. Also used as an antioxidant.

Solubility
Soluble in ethanol (50 mg/ml) and water (0.35 g/100 ml at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text