Thermo Scientific Chemicals

1-Iodo-3,5-bis(trifluoromethyl)benzene, 97+%, Thermo Scientific Chemicals

Catalog number: A10878.14
25 g, Each
Thermo Scientific Chemicals

1-Iodo-3,5-bis(trifluoromethyl)benzene, 97+%, Thermo Scientific Chemicals

Catalog number: A10878.14
25 g, Each
Quantity
Catalog number: A10878.14
also known as A10878-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
328-73-4
IUPAC Name
1-iodo-3,5-bis(trifluoromethyl)benzene
Molecular Formula
C8H3F6I
InChI Key
VDPIZIZDKPFXLI-UHFFFAOYSA-N
SMILES
FC(F)(F)C1=CC(=CC(I)=C1)C(F)(F)F
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥97.0%
Refractive Index
1.4620-1.4670 @ 20?C
Form
Liquid

Description

3,5-bis(trifluoromethyl)pheny1- magnesium iodide was prepared from 1-iodo-3,5- bis(trifluoromethyl)benzene. Monodeprotected masked hexayne was not particularly stable and hence was used directly without purification in a Sonogashira coupling with 1-iodo-3,5-bis(trifluoromethyl)benzene to provide mono-aryl-end-capped masked hexayne.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-bis(trifluoromethyl)pheny1- magnesium iodide was prepared from 1-iodo-3,5- bis(trifluoromethyl)benzene. Monodeprotected masked hexayne was not particularly stable and hence was used directly without purification in a Sonogashira coupling with 1-iodo-3,5-bis(trifluoromethyl)benzene to provide mono-aryl-end-capped masked hexayne.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text