Thermo Scientific Chemicals

Methyl 4-hydroxy-3-nitrobenzoate, 98+%, Thermo Scientific Chemicals

Catalog number: A10880.30
250 g, Each
Thermo Scientific Chemicals

Methyl 4-hydroxy-3-nitrobenzoate, 98+%, Thermo Scientific Chemicals

Catalog number: A10880.30
250 g, Each
Quantity
Catalog number: A10880.30
also known as A10880-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
99-42-3
IUPAC Name
methyl 4-hydroxy-3-nitrobenzoate
Molecular Formula
C8H7NO5
InChI Key
GNCWCTBHZCBXGL-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=C(O)C(=C1)[N+]([O-])=O
Assay (GC)
≥98.0%
Form
Powder
Melting Point (clear melt)
72.5-78.5?C
Appearance (Color)
Yellow

Description

4-alkoxy-3-nitrobenzoic acids were obtained by the alkylation of methyl 4-hydroxy-3-nitrobenzoate with corresponding n-alkylbromides in 3-pentanone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-alkoxy-3-nitrobenzoic acids were obtained by the alkylation of methyl 4-hydroxy-3-nitrobenzoate with corresponding n-alkylbromides in 3-pentanone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text