Thermo Scientific Chemicals

Methyl p-toluenesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: A10881.0B
1000 g, Each
Thermo Scientific Chemicals

Methyl p-toluenesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: A10881.0B
1000 g, Each
Quantity
Catalog number: A10881.0B
also known as A10881-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
80-48-8
IUPAC Name
methyl 4-methylbenzene-1-sulfonate
Molecular Formula
C8H10O3S
InChI Key
VUQUOGPMUUJORT-UHFFFAOYSA-N
SMILES
COS(=O)(=O)C1=CC=C(C)C=C1
Appearance (Color)
Colorless to pale yellow to yellow to brown
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder or clear liquid as melt
Refractive Index
1.5160-1.5200 @ 20?C

Description

Methyl p-toluenesulfonate is used as a methylating agent in organic synthesis. It acts as a catalyst for alkyd resins. It is also employed in selective 1-substitution reaction of tetrazole. Further, it is used in the preparation of dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl p-toluenesulfonate is used as a methylating agent in organic synthesis. It acts as a catalyst for alkyd resins. It is also employed in selective 1-substitution reaction of tetrazole. Further, it is used in the preparation of dyes.

Solubility
Soluble in ether, alcohol, benzene and chloroform. Slightly soluble in petroleum ether. Insoluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong acids and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text