Thermo Scientific Chemicals

Methanesulfonamide, 97+%, Thermo Scientific Chemicals

Catalog number: A10885.09
10 g, Each
Thermo Scientific Chemicals

Methanesulfonamide, 97+%, Thermo Scientific Chemicals

Catalog number: A10885.09
10 g, Each
Quantity
Catalog number: A10885.09
also known as A10885-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3144-09-0
Assay from Suppliers CofA
≥97.0%
Melting Point
85-93°C
Appearance (Color)
White to yellow to pale brown
Form
Crystals or crystalline powder or flakes

Description

Methanesulfonamide will react with thionyl chloride, cyanates, carbon disulfide and ketones and aldehydes to prepare pharmaceuticals, brightening agent and other target molecules. It is also used in biological studies to predict binding affinity and binding mode of protein ligand complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methanesulfonamide will react with thionyl chloride, cyanates, carbon disulfide and ketones and aldehydes to prepare pharmaceuticals, brightening agent and other target molecules. It is also used in biological studies to predict binding affinity and binding mode of protein ligand complexes.

Solubility
Slightly soluble in DMSO and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text