Antimony potassium tartrate hydrate, 98%, Thermo Scientific Chemicals
Antimony potassium tartrate hydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Antimony potassium tartrate hydrate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
100 g
500 g
Catalog number A10889.0E
also known as A10889-0E
Price (USD)/ Each
491.65
Online exclusive
546.00 
Save 54.35 (10%)
-
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (USD)/ Each
491.65
Online exclusive
546.00 
Save 54.35 (10%)
Add to cart
Antimony potassium tartrate hydrate, 98%, Thermo Scientific Chemicals
Catalog numberA10889.0E
Price (USD)/ Each
491.65
Online exclusive
546.00 
Save 54.35 (10%)
-
Add to cart
Chemical Identifiers
CAS331753-56-1
IUPAC Namedipotassium 5,11-dioxo-2,6,8,12,13,14-hexaoxa-1,7-distibatricyclo[8.2.1.1⁴,⁷]tetradecane-3,9-dicarboxylate
Molecular FormulaC8H4K2O12Sb2
InChI KeyGUJUCWZGYWASLH-UHFFFAOYNA-J
SMILES[K+].[K+].[O-]C(=O)C1O[Sb]2OC(C(O[Sb]3OC1C(=O)O3)C([O-])=O)C(=O)O2
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or lumps
Loss on Drying≤6.83% (200?C/constant weight)
Appearance (Color)White
Assay (Redox Titration)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Antimony potassium tartrate hydrate is a compound used in the treatment of schistosomiasis. It is also used in the textile industry to aid in binding certain dyes to fabrics. It finds application in medicine as an expectorant and a nauseant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Antimony potassium tartrate hydrate is a compound used in the treatment of schistosomiasis. It is also used in the textile industry to aid in binding certain dyes to fabrics. It finds application in medicine as an expectorant and a nauseant.

Solubility
Soluble in water. Insoluble in ethanol.

Notes
Incompatible with mineral acids, strong bases, carbonates, lead, silver salts and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Vagvala, T. C.; Pandey, S. S.; Ogomi, Y.; Ma, T.; Hayase, S. Investigation of metal xanthates as latent curing catalysts for epoxy resin via formation of in-situ metal sulfides. Inorg. Chim. Acta 2015, 435, 292-298.
  2. Nguyen, T. H.; Müller, R. H.; Taupitz, M.; Schnorr, J.; Hamm, B.; Wagner, S. Novel oral phosphate binder with nanocrystalline maghemite-phosphate binding capacity and pH effect. Int. J. Pharm. 2015, 482 (1), 21-26.