Thermo Scientific Chemicals

2-Amino-6-chlorobenzoic acid, 97+%, Thermo Scientific Chemicals

Catalog number: A10890.14
25 g, Each
Thermo Scientific Chemicals

2-Amino-6-chlorobenzoic acid, 97+%, Thermo Scientific Chemicals

Catalog number: A10890.14
25 g, Each
Quantity
Catalog number: A10890.14
also known as A10890-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2148-56-3
IUPAC Name
2-amino-6-chlorobenzoic acid
Molecular Formula
C7H6ClNO2
InChI Key
SZCPTRGBOVXVCA-UHFFFAOYSA-N
SMILES
NC1=CC=CC(Cl)=C1C(O)=O
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Form
Powder
Assay (HPLC)
≥97.5%
Appearance (Color)
Cream to pale brown

Description

2-Amino-6-chlorobenzoic acid is used as intermediate for medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-6-chlorobenzoic acid is used as intermediate for medicine.

Solubility
Very soluble in water. Soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text