Thermo Scientific Chemicals

4-Bromobenzyl bromide, 98+%, Thermo Scientific Chemicals

Catalog number: A10892.14
25 g, Each
Thermo Scientific Chemicals

4-Bromobenzyl bromide, 98+%, Thermo Scientific Chemicals

Catalog number: A10892.14
25 g, Each
Quantity
Catalog number: A10892.14
also known as A10892-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
589-15-1
IUPAC Name
1-bromo-4-(bromomethyl)benzene
Molecular Formula
C7H6Br2
InChI Key
YLRBJYMANQKEAW-UHFFFAOYSA-N
SMILES
BrCC1=CC=C(Br)C=C1
Appearance (Color)
White to pale cream
Assay from Supplier's CofA
≥98.0% (GC)
Form
Powder
Melting Point
62.0-64.0°C

Description

4-Bromobenzyl bromide is used to prepare a 20-member aminomethyl-substituted biaryl library via sequential N-alkylation of various amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzyl bromide is used to prepare a 20-member aminomethyl-substituted biaryl library via sequential N-alkylation of various amines.

Solubility
Easily soluble in hot alcohol, ether, carbon disulfide, benzene and glacial acetic acid. Soluble in water and alcohol.

Notes
Keep containers tightly closed in a dry, cool and well-ventilated place. Store at 4°C. Incompatible with strong oxidizing agents. It is sensitive to light.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text