Methyl m-toluate, 98%, Thermo Scientific Chemicals
Methyl m-toluate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl m-toluate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number A10893.18
also known as A10893-18
Price (USD)/ Each
39.70
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
39.70
Add to cart
Methyl m-toluate, 98%, Thermo Scientific Chemicals
Catalog numberA10893.18
Price (USD)/ Each
39.70
-
Add to cart
Chemical Identifiers
CAS99-36-5
IUPAC Namemethyl 3-methylbenzoate
Molecular FormulaC9H10O2
InChI KeyCPXCDEMFNPKOEF-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Refractive Index1.5140-1.5180 @ 20?C
FormLiquid
Methyl3-methylcyclohexanecarboxylate was prepared by hydrogenation of methyl m-toluate in acetic acid solvent over Adams catalyst. analysis for valproic acid (VA) in serum, for which we use a gas chromatograph equipped with a flame ionization detector, with methyl-m-toluate (MMT) as internal standard.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl3-methylcyclohexanecarboxylate was prepared by hydrogenation of methyl m-toluate in acetic acid solvent over Adams catalyst. analysis for valproic acid (VA) in serum, for which we use a gas chromatograph equipped with a flame ionization detector, with methyl-m-toluate (MMT) as internal standard.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Zeming Chen,; Yue Zhao,; Li-Min Wang. Enthalpy and dielectric relaxations in supercooled methyl m-toluate . J. Chem. Phys. 2009, 130 (20), 204515 .
  2. Tylinski, M.; Sepulveda, A.; Guiseppi-Elie, A.; Richert, R.; Chua, Y. Z.; Schick, C.; Ediger, M. D. title . APS. 2015,