Thermo Scientific Chemicals

Methyl m-toluate, 98%, Thermo Scientific Chemicals

Catalog number: A10893.30
250 g, Each
Thermo Scientific Chemicals

Methyl m-toluate, 98%, Thermo Scientific Chemicals

Catalog number: A10893.30
250 g, Each
Quantity
Catalog number: A10893.30
also known as A10893-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
99-36-5
IUPAC Name
methyl 3-methylbenzoate
Molecular Formula
C9H10O2
InChI Key
CPXCDEMFNPKOEF-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CC(C)=C1
Form
Liquid
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Refractive Index
1.5140-1.5180 @ 20?C

Description

Methyl3-methylcyclohexanecarboxylate was prepared by hydrogenation of methyl m-toluate in acetic acid solvent over Adams catalyst. analysis for valproic acid (VA) in serum, for which we use a gas chromatograph equipped with a flame ionization detector, with methyl-m-toluate (MMT) as internal standard.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl3-methylcyclohexanecarboxylate was prepared by hydrogenation of methyl m-toluate in acetic acid solvent over Adams catalyst. analysis for valproic acid (VA) in serum, for which we use a gas chromatograph equipped with a flame ionization detector, with methyl-m-toluate (MMT) as internal standard.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text