Thermo Scientific Chemicals

Boric acid, 99+%, Thermo Scientific Chemicals

Catalog number: A10896.0C
10,000 g, Each
Thermo Scientific Chemicals

Boric acid, 99+%, Thermo Scientific Chemicals

Catalog number: A10896.0C
10,000 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A10896.0C
also known as A10896-0C
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10043-35-3
IUPAC Name
boric acid
Molecular Formula
BH3O3
InChI Key
KGBXLFKZBHKPEV-UHFFFAOYSA-N
SMILES
OB(O)O
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥99.0 to ≤101.0%
Comment
Material sourced in the U.S. and in other countries
Identification (FTIR)
Conforms (non-U.S. sourced material)

Description

Boric acid is a precursor material for other boron compounds. A dilute water solution of boric acid is usually employed as a mild antiseptic and eyewash. Boric acid is employed in leather manufacture, electroplating and cosmetics. It is involved in the production of monofilament fiberglass which finds applications in boats, industrial piping, LCD flat panel displays and computer circuit boards. It is in combination with sodium tetraborate decahydrate (borax) and is used as a welding flux by blacksmiths. The mixture of boric acid and silicone oil is useful in the production of silly putty.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Boric acid is a precursor material for other boron compounds. A dilute water solution of boric acid is usually employed as a mild antiseptic and eyewash. Boric acid is employed in leather manufacture, electroplating and cosmetics. It is involved in the production of monofilament fiberglass which finds applications in boats, industrial piping, LCD flat panel displays and computer circuit boards. It is in combination with sodium tetraborate decahydrate (borax) and is used as a welding flux by blacksmiths. The mixture of boric acid and silicone oil is useful in the production of silly putty.

Solubility
Soluble in water and alcohols. Slightly soluble in acetone and pyridine.

Notes
Store in cool place. Moisture sensitive. Incompatible with potassium and acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text