Thermo Scientific Chemicals

2-Ethylhexane-1,3-diol, 97%, Thermo Scientific Chemicals

Catalog number: A10898.0F
2500 mL, Each
Thermo Scientific Chemicals

2-Ethylhexane-1,3-diol, 97%, Thermo Scientific Chemicals

Catalog number: A10898.0F
2500 mL, Each
Quantity
Catalog number: A10898.0F
also known as A10898-0F
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
94-96-2
IUPAC Name
2-ethylhexane-1,3-diol
Molecular Formula
C8H18O2
InChI Key
RWLALWYNXFYRGW-UHFFFAOYNA-N
SMILES
CCCC(O)C(CC)CO
Form
Liquid
Refractive Index
1.4485-1.4535 @ 20?C
Appearance (Color)
Clear colorless
Assay (Silylated GC)
≥96.0%
Identification (FTIR)
Conforms

Description

2-Ethylhexane-1,3-diol is used in organic synthesis and can be used to produce polyester resin, polyester plasticizers, polyurethane resin. It is also effective vermifuge to mosquitoes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylhexane-1,3-diol is used in organic synthesis and can be used to produce polyester resin, polyester plasticizers, polyurethane resin. It is also effective vermifuge to mosquitoes.

Solubility
Soluble in water (42g/L at 20°C). Soluble in alcohol and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is hygroscopic in nature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text