Thermo Scientific Chemicals

Aluminum potassium sulfate dodecahydrate, 98+%, Thermo Scientific Chemicals

Catalog number: A10906.0E
2500 g, Each
Thermo Scientific Chemicals

Aluminum potassium sulfate dodecahydrate, 98+%, Thermo Scientific Chemicals

Catalog number: A10906.0E
2500 g, Each
Quantity
Catalog number: A10906.0E
also known as A10906-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7784-24-9
IUPAC Name
aluminium(3+) potassium dodecahydrate disulfate
Molecular Formula
AlH24KO20S2
InChI Key
GNHOJBNSNUXZQA-UHFFFAOYSA-J
SMILES
O.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[K+].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Assay (unspecified)
≥98.0%
Identification (FTIR)
Conforms
Appearance (Color)
White or colorless translucent
Solution Test
5% in water: clear colorless
Form
Crystals or powder or crystalline powder or granules

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text