Aluminum potassium sulfate dodecahydrate, 98+%, Thermo Scientific Chemicals
Aluminum potassium sulfate dodecahydrate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum potassium sulfate dodecahydrate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2500 g
Catalog number A10906.36
also known as A10906-36
Price (USD)/ Each
85.65
Online exclusive
94.80 
Save 9.15 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
85.65
Online exclusive
94.80 
Save 9.15 (10%)
Add to cart
Aluminum potassium sulfate dodecahydrate, 98+%, Thermo Scientific Chemicals
Catalog numberA10906.36
Price (USD)/ Each
85.65
Online exclusive
94.80 
Save 9.15 (10%)
-
Add to cart
Chemical Identifiers
CAS7784-24-9
IUPAC Namealuminium(3+) potassium dodecahydrate disulfate
Molecular FormulaAlH24KO20S2
InChI KeyGNHOJBNSNUXZQA-UHFFFAOYSA-J
SMILESO.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[K+].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)≥98.0%
Identification (FTIR)Conforms
Appearance (Color)White or colorless translucent
Solution Test5% in water: clear colorless
FormCrystals or powder or crystalline powder or granules
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only