Thermo Scientific Chemicals

2,2'-Biquinoline, 98%, Thermo Scientific Chemicals

Catalog number: A10907.06
5 g, Each
Thermo Scientific Chemicals

2,2'-Biquinoline, 98%, Thermo Scientific Chemicals

Catalog number: A10907.06
5 g, Each
Quantity
Catalog number: A10907.06
also known as A10907-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
119-91-5
IUPAC Name
2,2'-biquinoline
Molecular Formula
C18H12N2
InChI Key
WPTCSQBWLUUYDV-UHFFFAOYSA-N
SMILES
C1=CC2=CC=C(N=C2C=C1)C1=NC2=CC=CC=C2C=C1
Form
Crystals or powder or crystalline powder or flakes
Assay (Non-aqueous acid-base Titration)
≥98.0 to ≤102.0%
Melting Point (clear melt)
192.0-199.0?C
Appearance (Color)
White to cream to pale brown

Description

2,2'-Biquinoline is used as a reagent for spectrophotometric determination of Cu and as an indicator for titration of organolithium reagents, giving yellow-green charge-transfer complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′-Biquinoline is used as a reagent for spectrophotometric determination of Cu and as an indicator for titration of organolithium reagents, giving yellow-green charge-transfer complexes.

Solubility
Sparingly soluble in water (0.001g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text