Thermo Scientific Chemicals

n-Butylbenzene, 99%, Thermo Scientific Chemicals

Catalog number: A10910.0B
1000 g, Each
Thermo Scientific Chemicals

n-Butylbenzene, 99%, Thermo Scientific Chemicals

Catalog number: A10910.0B
1000 g, Each
Quantity
Catalog number: A10910.0B
also known as A10910-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
104-51-8
IUPAC Name
butylbenzene
Molecular Formula
C10H14
InChI Key
OCKPCBLVNKHBMX-UHFFFAOYSA-N
SMILES
CCCCC1=CC=CC=C1
Assay (GC)
≥98.5%
Appearance (Color)
Clear colorless
Form
Liquid
Refractive Index
1.4875-1.4905 @ 20?C

Description

n-Butylbenzene is an organic solvent that has been used to induce cell death in vitro and for bioconversion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Butylbenzene is an organic solvent that has been used to induce cell death in vitro and for bioconversion.

Solubility
Sparingly soluble in water (0.012g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Storage temperature 2 - 8°C. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text