n-Butylbenzene, 99%, Thermo Scientific Chemicals
n-Butylbenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Butylbenzene, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1000 g
Catalog number A10910.30
also known as A10910-30
Price (USD)/ Each
153.65
Online exclusive
171.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
153.65
Online exclusive
171.00 
Save 17.35 (10%)
Add to cart
n-Butylbenzene, 99%, Thermo Scientific Chemicals
Catalog numberA10910.30
Price (USD)/ Each
153.65
Online exclusive
171.00 
Save 17.35 (10%)
-
Add to cart
Chemical Identifiers
CAS104-51-8
IUPAC Namebutylbenzene
Molecular FormulaC10H14
InChI KeyOCKPCBLVNKHBMX-UHFFFAOYSA-N
SMILESCCCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.4875-1.4905 @ 20?C
n-Butylbenzene is an organic solvent that has been used to induce cell death in vitro and for bioconversion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Butylbenzene is an organic solvent that has been used to induce cell death in vitro and for bioconversion.

Solubility
Sparingly soluble in water (0.012g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Storage temperature 2 - 8°C. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. A Roubaud.;, O Lemaire.;, R Minetti.; L.R Sochet. High pressure auto-ignition and oxidation mechanisms of o-xylene, o-ethyltoluene, and n-butylbenzene between 600 and 900 K. Combustion and Flame. 2000, 123, (4), 561-571.
  2. J. Alex G. Harrison.; Margaret S. Lin. Energy dependence of the fragmentation of the n-butylbenzene molecular ion. International Journal of Mass Spectrometry and Ion Physics. 1983, 52, (2-3), 353-356.