Thermo Scientific Chemicals

Potassium hexafluorotitanate, 97%, Thermo Scientific Chemicals

Catalog number: A10915.0E
2500 g, Each
Thermo Scientific Chemicals

Potassium hexafluorotitanate, 97%, Thermo Scientific Chemicals

Catalog number: A10915.0E
2500 g, Each
Quantity
Catalog number: A10915.0E
also known as A10915-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16919-27-0
IUPAC Name
titanium(4+) dipotassium hexafluoride
Molecular Formula
F6K2Ti
InChI Key
RXCBCUJUGULOGC-UHFFFAOYSA-H
SMILES
[F-].[F-].[F-].[F-].[F-].[F-].[K+].[K+].[Ti+4]
Form
powder
Appearance (Color)
White
Elemental Analysis
Potassium K : 31.27 - 33.87% (Theory 32.56%)
Elemental Analysis
Titanium Ti: 19.14 - 20.73% (Theory 19.94%)

Description

Potassium hexafluorotitanate is used as analytical reagent, alloying additive, flame retardant and flux agent for casting. It is also used in the manufacture of titanium acid and aluminum alloys containing titanium. It is also employed in metal surface treatment and finishing, detergent applications and dental impression molding. Further, it is used as a catalyst for polypropylene synthesis. In addition to this, it is used in the extraction-refining-processing of metals, electrical-electronics engineering, textile processing and in ceramics-glass industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium hexafluorotitanate is used as analytical reagent, alloying additive, flame retardant and flux agent for casting. It is also used in the manufacture of titanium acid and aluminum alloys containing titanium. It is also employed in metal surface treatment and finishing, detergent applications and dental impression molding. Further, it is used as a catalyst for polypropylene synthesis. In addition to this, it is used in the extraction-refining-processing of metals, electrical-electronics engineering, textile processing and in ceramics-glass industries.

Solubility
Soluble in hot water. Slightly soluble in cold water and inorganic acid. Insoluble in ammonia.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text