Thermo Scientific Chemicals

Sodium 1-heptanesulfonate, 99% (dry wt.), water <2%, Thermo Scientific Chemicals

Catalog number: A10917.06
5 g, Each
Thermo Scientific Chemicals

Sodium 1-heptanesulfonate, 99% (dry wt.), water <2%, Thermo Scientific Chemicals

Catalog number: A10917.06
5 g, Each
Quantity
Catalog number: A10917.06
also known as A10917-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
22767-50-6
IUPAC Name
sodium heptane-1-sulfonate
Molecular Formula
C7H15NaO3S
InChI Key
REFMEZARFCPESH-UHFFFAOYSA-M
SMILES
[Na+].CCCCCCCS([O-])(=O)=O
Assay (ex Sulphated Ash)
≥98.5 to ≤101.5% (dry wt. basis)
Loss on Drying
≤2.0% (105?C/constant wt.)
Appearance (Color)
White
Sulfated Ash
34.59-35.64 (Theory 35.12%)
Identification (FTIR)
Conforms

Description

HPLC reagent used for ion-association in analyses of peptides and proteinsSodium 1-heptanesulfonate is used as a ion-associating reagent for HPLC and involved in the analysis of peptides and proteins. It is utilized in the analysis of pharmaceutical products, organic and inorganic small molecule compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
HPLC reagent used for ion-association in analyses of peptides and proteinsSodium 1-heptanesulfonate is used as a ion-associating reagent for HPLC and involved in the analysis of peptides and proteins. It is utilized in the analysis of pharmaceutical products, organic and inorganic small molecule compounds.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text