Thermo Scientific Chemicals

Glycerol tripalmitate, 98%, Thermo Scientific Chemicals

Catalog number: A10922.14
25 g, Each
Thermo Scientific Chemicals

Glycerol tripalmitate, 98%, Thermo Scientific Chemicals

Catalog number: A10922.14
25 g, Each
Quantity
Catalog number: A10922.14
also known as A10922-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
555-44-2
IUPAC Name
1,3-bis(hexadecanoyloxy)propan-2-yl hexadecanoate
Molecular Formula
C51H98O6
InChI Key
PVNIQBQSYATKKL-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCC
Appearance (Color)
White to pale cream
Form
Powder
Assay (HPLC)
≥97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text