Thermo Scientific Chemicals

4-tert-Butylaniline, 98+%, Thermo Scientific Chemicals

Catalog number: A10925.22
100 g, Each
Thermo Scientific Chemicals

4-tert-Butylaniline, 98+%, Thermo Scientific Chemicals

Catalog number: A10925.22
100 g, Each
Quantity
Catalog number: A10925.22
also known as A10925-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
769-92-6
IUPAC Name
4-tert-butylaniline
Molecular Formula
C10H15N
InChI Key
WRDWWAVNELMWAM-UHFFFAOYSA-N
SMILES
CC(C)(C)C1=CC=C(N)C=C1
Form
Liquid
Refractive Index
1.5365-1.5395 @ 20°C
Assay (GC)
≥98.0%
Appearance (Color)
Clear colorless to pale yellow to yellow to orange or pale pink
Identification (FTIR)
Conforms

Description

4-tert-Butylaniline reacts with formic acid to produce formic acid-(4-tert-butyl-anilide). This reaction could happen in the presence of the solvent of toluene in the condition of heating. 4-tert-Butylaniline is used in the synthesis of 4-tert-Butyl-4?,4?-dinitrotriphenylamine, new triphenylamine-containing diamine monomer and 2-oxopyrimido[4,5-d]pyrimidin-5(6H)-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butylaniline reacts with formic acid to produce formic acid-(4-tert-butyl-anilide). This reaction could happen in the presence of the solvent of toluene in the condition of heating. 4-tert-Butylaniline is used in the synthesis of 4-tert-Butyl-4′,4″-dinitrotriphenylamine, new triphenylamine-containing diamine monomer and 2-oxopyrimido[4,5-d]pyrimidin-5(6H)-one.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive to light. Incompatible with strong acids, acid chlorides, acid anhydrides and chloroformates.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text