3-Fluoroanisole, 99%, Thermo Scientific Chemicals
3-Fluoroanisole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Fluoroanisole, 99%, Thermo Scientific Chemicals

Have Questions?
Quantity:
10 g
Catalog number A10931.09
also known as A10931-09
Price (USD)/ Each
28.50
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
28.50
Add to cart
3-Fluoroanisole, 99%, Thermo Scientific Chemicals
Catalog numberA10931.09
Price (USD)/ Each
28.50
-
Add to cart
Chemical Identifiers
CAS456-49-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow or pale orange
FormLiquid
Assay (GC)≥98.5%
CommentSpecification differs for U.S. and non-U.S. material where indicated
Identification (FTIR)Conforms [non-U.S. specification]
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only