Thermo Scientific Chemicals

1-Butyl isothiocyanate, 98+%, Thermo Scientific Chemicals

Catalog number: A10935.22
100 g, Each
Thermo Scientific Chemicals

1-Butyl isothiocyanate, 98+%, Thermo Scientific Chemicals

Catalog number: A10935.22
100 g, Each
Quantity
Catalog number: A10935.22
also known as A10935-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
592-82-5
IUPAC Name
1-isothiocyanatobutane
Molecular Formula
C5H9NS
InChI Key
LIMQQADUEULBSO-UHFFFAOYSA-N
SMILES
CCCCN=C=S
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥98.0%
Refractive Index
1.4985-1.5025 @ 20?C

Description

1-Butyl isothiocyanate participates in the synthesis and characterization of Poly(N-Propargylthiourea). Participates in the Synthesis of 2-Aminobenzothiazoles via Copper (I)-Catalyzed Cross-Coupling with Part-Per-Million Catalyst Loadings. Thiols reactions with 1-Butyl isothiocyanate in aquatic toxicity data in the TETRATOX assay and reactivity data in an abiotic thiol assay. utyl isothiocyanate has been used as capping reagent to immobilize the functionalized derivative of Oregon Green 514 molecular switch on a preformed amino self assembled monlayer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Butyl isothiocyanate participates in the synthesis and characterization of Poly(N-Propargylthiourea). Participates in the Synthesis of 2-Aminobenzothiazoles via Copper (I)-Catalyzed Cross-Coupling with Part-Per-Million Catalyst Loadings. Thiols reactions with 1-Butyl isothiocyanate in aquatic toxicity data in the TETRATOX assay and reactivity data in an abiotic thiol assay. utyl isothiocyanate has been used as capping reagent to immobilize the functionalized derivative of Oregon Green 514 molecular switch on a preformed amino self assembled monlayer.

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text