Thermo Scientific Chemicals

Acetaldehyde dimethyl acetal, 98+%, Thermo Scientific Chemicals

Catalog number: A10939.36
500 g, Each
Thermo Scientific Chemicals

Acetaldehyde dimethyl acetal, 98+%, Thermo Scientific Chemicals

Catalog number: A10939.36
500 g, Each
Quantity
Catalog number: A10939.36
also known as A10939-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
534-15-6
IUPAC Name
1,1-dimethoxyethane
Molecular Formula
C4H10O2
InChI Key
SPEUIVXLLWOEMJ-UHFFFAOYSA-N
SMILES
COC(C)OC
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless
Form
Liquid
Refractive Index
1.3650-1.3690 @ 20?C

Description

Acetaldehyde dimethyl acetal may be used in the preparation of glucoside derivatives of steganol. It may be used as polymer solvent for the encapsulation of water-soluble model protein, bovine serum albumin into biodegradable poly(D,L-lactic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetaldehyde dimethyl acetal may be used in the preparation of glucoside derivatives of steganol. It may be used as polymer solvent for the encapsulation of water-soluble model protein, bovine serum albumin into biodegradable poly(D,L-lactic acid.

Solubility
Soluble in water (1000 mg/ml at 25°C), alcohols, chloroform, ether, and acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text