Thermo Scientific Chemicals

p-Anisidine, 99%, Thermo Scientific Chemicals

Catalog number: A10946.22
100 g, Each
Thermo Scientific Chemicals

p-Anisidine, 99%, Thermo Scientific Chemicals

Catalog number: A10946.22
100 g, Each
Quantity
Catalog number: A10946.22
also known as A10946-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
104-94-9
IUPAC Name
4-methoxyaniline
Molecular Formula
C7H9NO
InChI Key
BHAAPTBBJKJZER-UHFFFAOYSA-N
SMILES
COC1=CC=C(N)C=C1
Melting Point (clear melt)
56-61?C
Form
Crystals or powder or crystalline powder or flakes or granules
Identification (FTIR)
Conforms
Appearance (Color)
Cream to dark brown or grey or grey/brown
Assay (GC)
≥98.5%

Description

p-Anisidine is used in the production of azo dyes. It is also used for biochemical research and in organic synthesis. Further, it is used for detecting unsaturated aldehydes. It is also employed in food quality testing. In addition to this, it is used in the preparation of 1-methoxy-4-trans-styryl-benzene by reacting with vinylbenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Anisidine is used in the production of azo dyes. It is also used for biochemical research and in organic synthesis. Further, it is used for detecting unsaturated aldehydes. It is also employed in food quality testing. In addition to this, it is used in the preparation of 1-methoxy-4-trans-styryl-benzene by reacting with vinylbenzene.

Solubility
Soluble in acetone, benzene, ether and ethanol. Insoluble in water.

Notes
Air and light sensitive. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides and chloroformates.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text