Thermo Scientific Chemicals

p-Toluoylacetonitrile, 97%, Thermo Scientific Chemicals

Catalog number: A10950.03
1 g, Each
Thermo Scientific Chemicals

p-Toluoylacetonitrile, 97%, Thermo Scientific Chemicals

Catalog number: A10950.03
1 g, Each
Quantity
Catalog number: A10950.03
also known as A10950-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7391-28-8
IUPAC Name
3-(4-methylphenyl)-3-oxopropanenitrile
Molecular Formula
C10H9NO
InChI Key
AIECDYDQPCANJK-UHFFFAOYSA-N
SMILES
CC1=CC=C(C=C1)C(=O)CC#N
Appearance (Color)
Pale cream to cream
Melting Point (clear melt)
99.0-108.0?C
Form
Powder or needles or flakes or plates
Assay (GC)
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text