Thermo Scientific Chemicals

3-Methoxyphenylacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: A10951.22
100 g, Each
Thermo Scientific Chemicals

3-Methoxyphenylacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: A10951.22
100 g, Each
Quantity
Catalog number: A10951.22
also known as A10951-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1798-09-0
IUPAC Name
2-(3-methoxyphenyl)acetate
Molecular Formula
C9H9O3
InChI Key
LEGPZHPSIPPYIO-UHFFFAOYSA-M
SMILES
COC1=CC=CC(CC([O-])=O)=C1
Appearance (Color)
White to cream to pale brown
Form
Crystals or powder or crystalline powder and/or chunks
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Assay (Silylated GC)
≥96.0%
Melting Point
64.0-73.0°C

Description

A fluorimetric method for the estimation of 4-hydroxy-3-methoxyphenylacetic acid (homovanillic acid) has been developed and applied to normal brain tissue. The presence of homovanillic acid in the caudate nucleus of normal animals of several species has been demonstrated.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A fluorimetric method for the estimation of 4-hydroxy-3-methoxyphenylacetic acid (homovanillic acid) has been developed and applied to normal brain tissue. The presence of homovanillic acid in the caudate nucleus of normal animals of several species has been demonstrated.

Solubility
Soluble in Chloroform and Ethyl Acetate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text