Thermo Scientific Chemicals

1,4-Naphthoquinone, 97+% (dry wt.), cont. up to 5% water, Thermo Scientific Chemicals

Catalog number: A10958.0E
2500 g, Each
Thermo Scientific Chemicals

1,4-Naphthoquinone, 97+% (dry wt.), cont. up to 5% water, Thermo Scientific Chemicals

Catalog number: A10958.0E
2500 g, Each
Quantity
Catalog number: A10958.0E
also known as A10958-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
130-15-4
IUPAC Name
1,4-dihydronaphthalene-1,4-dione
Molecular Formula
C10H6O2
InChI Key
FRASJONUBLZVQX-UHFFFAOYSA-N
SMILES
O=C1C=CC(=O)C2=CC=CC=C12
Form
Powder
Appearance (Color)
Dark yellow to yellow to brown
Assay (GC)
≥97.0%
Water Content (Karl Fischer Titration)
≤5%
Identification (FTIR)
Conforms

Description

1,4-Naphthoquinone is used as a strong dienophile in Diels-Alder reaction. It is also used as a precursor to anthroquinone and 5-nitro-1,4-naphthalenedione, which finds application in the preparation of aminoanthroquinone and is also used as a dye precursor. It is a basic component of Vitamin K.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Naphthoquinone is used as a strong dienophile in Diels-Alder reaction. It is also used as a precursor to anthroquinone and 5-nitro-1,4-naphthalenedione, which finds application in the preparation of aminoanthroquinone and is also used as a dye precursor. It is a basic component of Vitamin K.

Solubility
Soluble in alkali hydroxide solvents, sparingly soluble in petroleum ether and water. Freely soluble in carbondioxide, acetic acid, benzene and choloroform.

Notes
Light sensitive. Incompatible with strong oxidizing agents and strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text