Thermo Scientific Chemicals

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide, 98%, Thermo Scientific Chemicals

Catalog number: A10962.06
5 g, Each
Thermo Scientific Chemicals

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide, 98%, Thermo Scientific Chemicals

Catalog number: A10962.06
5 g, Each
Quantity
Catalog number: A10962.06
also known as A10962-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
22572-40-3
IUPAC Name
(3-{[(ethylimino)methylidene]amino}propyl)trimethylazanium iodide
Molecular Formula
C9H20IN3
InChI Key
AGSKWMRPXWHSPF-UHFFFAOYSA-M
SMILES
[I-].CCN=C=NCCC[N+](C)(C)C
Appearance (Color)
White to cream to yellow
Form
Crystalline powder or powder
Assay (Titration ex Iodide)
≥97.5 to ≤102.5%
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
≤1.0%

Description

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide is in proteins as water soluble carboxyl modifying reagent. It is used as a crosslinking agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide is in proteins as water soluble carboxyl modifying reagent. It is used as a crosslinking agent.

Solubility
Soluble in water.

Notes
Keep in a dry, cool and well-ventilated place. Keep container tightly closed. Incompatible with acids, strong oxidizing agents, moist air and water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text