Thermo Scientific Chemicals

2,4,6-Trifluoroaniline, 98%, Thermo Scientific Chemicals

Catalog number: A10964.14
25 g, Each
Thermo Scientific Chemicals

2,4,6-Trifluoroaniline, 98%, Thermo Scientific Chemicals

Catalog number: A10964.14
25 g, Each
Quantity
Catalog number: A10964.14
also known as A10964-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
363-81-5
IUPAC Name
2,4,6-trifluoroaniline
Molecular Formula
C6H4F3N
InChI Key
BJSVKBGQDHUBHZ-UHFFFAOYSA-N
SMILES
NC1=C(F)C=C(F)C=C1F
Form
Crystals or powder or crystalline powder or fused solid
Melting Point (clear melt)
31.0-37.0?C
Assay (GC)
≥97.5%
Appearance (Color)
White to pale yellow

Description

2,4,6-Trifluoroaniline was used in the synthesis of 3-nitro-2,4,6 -trifluoroacetanilide; eries of N?-phenyl-N-(1- phenyl cyclopentyl)-methyl ureas and 4-substituted 2,6-difluoro N-aryl pyridinones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trifluoroaniline was used in the synthesis of 3-nitro-2,4,6 -trifluoroacetanilide; eries of N′-phenyl-N-(1- phenyl cyclopentyl)-methyl ureas and 4-substituted 2,6-difluoro N-aryl pyridinones.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text