Invitrogen™

BioPrime™ Total FFPE Genomic Labeling System

Catalog number: A10965011
Have Questions?
Invitrogen™

BioPrime™ Total FFPE Genomic Labeling System

Catalog number: A10965011
Have Questions?
Catalog Number
A10965011
also known as A10965-011
Unit Size
30 rxns
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A10965011
also known as A10965-011
30 rxns
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The BioPrime™ Total FFPE Genomic Labeling System is a complete genomic DNA labeling kit designed for use in array comparative genomic hybridization applications (aCGH) using formalin-fixed paraffin-embedded (FFPE) samples. Previous protocols have required large amounts of very high quality DNA, which is often difficult to obtain from FFPE material. With a random prime amplification (RPA) method, using labeling mixes specifically formulated for FFPE samples, BioPrime™ Total FFPE Genomic Labeling System generates high yields of labeled DNA which is most representative of the starting sample.. Increased call accuracy is achieved with higher yields of labeled material on the array, giving you more accurate calls. The BioPrime™ Total FFPE Genomic Labeling System offers users the following advantages:

- Improved call rate with higher signal to noise ratios from Alexa Fluor™ 3 and 5 dyes
- Easy cleanup with PureLink™ purification eliminates the need for complicated volume-reduction steps
- Less channel bias due to enhanced reaction formulation for FFPE samples
- Lower requirements for input material using enzymatic RPA method


This complete, all-inclusive kit is optimized to work across a wider range of sample input material with no need for pre-amplification. Highly concentrated exo-Klenow fragment of DNA polymerase I in a component limited reaction allow for consistent, robust DNA yields of genomic FFPE DNA input. BioPrime™ Total for Agilent™ aCGH is supplied with Purelink™ purification which allows for low elution, eliminating the need for complicated volume-reduction steps. Purelink™ purification also reduces background due to dye bled found in other purification systems An optimized dye labeled nucleotide mix with new dNTP linkers, novel, application-specific Alexa Fluor™ 3 and 5 dyes, and improved buffer chemistry reduce labeling variation and increase signal to noise on arrays. Excitation and emission spectra of Alexa Fluor™ 3 and 5 dyes are suited for conventional two color scanners with no need to change settings. Complete kits are batch tested to assure quality in performance from lot to lot.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Fluorescence
Label or Dye
Alexa Fluor 3, Alexa Fluor 5
Final Product Type
Probes (Labeled DNA)
Sample Type
Tissue (FFPE)
Labeling Target
Genomic DNA
Excitation/Emission
650/670, 555/565
Includes Label or Dye
Yes
Labeling Method
Direct Labeling
No. of Reactions
30
Product Line
BioPrime™
Product Type
Genomic Labeling System
Quantity
30 Reactions

Contents & Storage

Contains Alexa Fluor™ 3 FFPE 10X nucleotide mix and Alexa Fluor™ 5 FFPE 10X Nucleotide Mix, Exo-Klenow fragment (40 U⁄μl), 5 mM EDTA, 2.5X Random Primers solution, control DNA (Salmon Sperm), and DEPC-treated water. Store at -80°C for long term storage. The 10X Nucleotide Mixes may be stored at +4°C for up to 4 weeks and should be protected from light. Also contains Purelink™ Genomic DNA purification columns and purification buffers. Store at room temperature. All components are guaranteed for 6 months when properly stored.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text