Thermo Scientific Chemicals

(R)-(+)-1-Phenylethylamine, 98%, Thermo Scientific Chemicals

Catalog number: A10967.22
100 g, Each
Thermo Scientific Chemicals

(R)-(+)-1-Phenylethylamine, 98%, Thermo Scientific Chemicals

Catalog number: A10967.22
100 g, Each
Quantity
Catalog number: A10967.22
also known as A10967-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3886-69-9
IUPAC Name
1-phenylethan-1-amine
Molecular Formula
C8H11N
InChI Key
RQEUFEKYXDPUSK-UHFFFAOYNA-N
SMILES
CC(N)C1=CC=CC=C1
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥97.5%
Optical Rotation
29.0 ± 1.5? (C=10 in ethanol)
Form
Liquid

Description

(R)-(+)-1-Phenylethylamine is a chiral amine used for the determination of the enantiomeric purity of acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-1-Phenylethylamine is a chiral amine used for the determination of the enantiomeric purity of acids.

Solubility
Soluble in water (40 g/L).

Notes
Air sensitive. Incompatible with oxidizing agents, acids, acid chlorides and acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text