Thermo Scientific Chemicals

Furfuryl alcohol, 98%, Thermo Scientific Chemicals

Catalog number: A10968.18
50 g, Each
Thermo Scientific Chemicals

Furfuryl alcohol, 98%, Thermo Scientific Chemicals

Catalog number: A10968.18
50 g, Each
Quantity
Catalog number: A10968.18
also known as A10968-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
98-00-0
IUPAC Name
(furan-2-yl)methanol
Molecular Formula
C5H6O2
InChI Key
XPFVYQJUAUNWIW-UHFFFAOYSA-N
SMILES
OCC1=CC=CO1
Refractive Index
1.4850-1.4890 @ 20?C
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥97.5%
Form
Liquid
Identification (FTIR)
Conforms

Description

Furfuryl alcohol was used in the synthesis of continuous titanium carbide nanofibers/nanoribbon. Furfuryl alcohol is an efficient trapping agent for singlet oxygen determination in natural waters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Furfuryl alcohol was used in the synthesis of continuous titanium carbide nanofibers/nanoribbon. Furfuryl alcohol is an efficient trapping agent for singlet oxygen determination in natural waters.

Solubility
Soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep containers tightly closed in a cool, well-ventilated place. Keep in properly labeled containers. Store at room temperature. Incompatible with strong oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text