Thermo Scientific Chemicals

Acetic hydrazide, 96%, Thermo Scientific Chemicals

Catalog number: A10969.14
25 g, Each
Thermo Scientific Chemicals

Acetic hydrazide, 96%, Thermo Scientific Chemicals

Catalog number: A10969.14
25 g, Each
Quantity
Catalog number: A10969.14
also known as A10969-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1068-57-1
IUPAC Name
acetohydrazide
Molecular Formula
C2H6N2O
InChI Key
OFLXLNCGODUUOT-UHFFFAOYSA-N
SMILES
CC(=O)NN
Form
Crystals or crystalline powder or chunks/lumps
Comment
Material sourced in the U.S. and in other countries
Melting Point (clear melt)
60-68°C
Appearance (Color)
Translucent colorless or white
Assay (unspecified)
≥95.0%

Description

Acetic hydrazide is used as an antibiotic for the treatment of mycobacterium tuberculosis. It is also involved in the preparation of sunitinib b reacting with 5-fluoroisatin. Further, it inhibits mycolic acid biosynthesis. It serves as an important intermediate in organic synthesis such as nitric furosemide Long (nifuratrone) in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetic hydrazide is used as an antibiotic for the treatment of mycobacterium tuberculosis. It is also involved in the preparation of sunitinib b reacting with 5-fluoroisatin. Further, it inhibits mycolic acid biosynthesis. It serves as an important intermediate in organic synthesis such as nitric furosemide Long (nifuratrone) in medicine.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text