Thermo Scientific Chemicals

N,N'-Dicyclohexylcarbodiimide, 99%, Thermo Scientific Chemicals

Catalog number: A10973.14
25 g, Each
Thermo Scientific Chemicals

N,N'-Dicyclohexylcarbodiimide, 99%, Thermo Scientific Chemicals

Catalog number: A10973.14
25 g, Each
Quantity
Catalog number: A10973.14
also known as A10973-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
538-75-0
IUPAC Name
N,N'-dicyclohexylmethanediimine
Molecular Formula
C13H22N2
InChI Key
QOSSAOTZNIDXMA-UHFFFAOYSA-N
SMILES
C1CCC(CC1)N=C=NC1CCCCC1
Appearance (Color)
White to pale yellow
Melting Point
32-36?C
Form
Crystalline mass or fused or waxy solid
Assay (GC)
≥98.5%

Description

N,N'-Dicyclohexylcarbodiimide is a carboxyl activating reagent used in peptide synthesis. It acts as a dehydrating agent for the preparation of amides, ketones and nitriles. It is involved in the preparation of esters by reacting tertiary alcohols with carboxylic acid. It is utilized for the stereo chemical inversion of secondary alcohols by forming formyl ester followed by saponification. It is associated with dimethyl sulfoxide and used in Pfitzner-Moffatt oxidation to convert alcohols into aldehydes and ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Dicyclohexylcarbodiimide is a carboxyl activating reagent used in peptide synthesis. It acts as a dehydrating agent for the preparation of amides, ketones and nitriles. It is involved in the preparation of esters by reacting tertiary alcohols with carboxylic acid. It is utilized for the stereo chemical inversion of secondary alcohols by forming formyl ester followed by saponification. It is associated with dimethyl sulfoxide and used in Pfitzner-Moffatt oxidation to convert alcohols into aldehydes and ketones.

Solubility
Soluble in methanol, chloroform, dichloromethane, acetonitrile, tetrahydrofuran and dimethylformamide. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text