Thermo Scientific Chemicals

Triethyl 4-phosphonocrotonate, cis + trans, 94%, Thermo Scientific Chemicals

Catalog number: A10995.14
25 g, Each
Thermo Scientific Chemicals

Triethyl 4-phosphonocrotonate, cis + trans, 94%, Thermo Scientific Chemicals

Catalog number: A10995.14
25 g, Each
Quantity
Catalog number: A10995.14
also known as A10995-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10236-14-3
Form
Liquid
Refractive Index
1.4510-1.4580 @ 20?C
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥92.0%

Description

Triethyl 4-phosphonocrotonate is used as a reactant for iminodipropionic acid as a leaving group for DNA polymerization by HIV-1 reverse transcriptase, orally bioavailable GPR40 agonist synthesis, preparation of mGlu4R agonists and synthesis of fluoroketone inhibitors of group VIA calcium-independent phospholipase A2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyl 4-phosphonocrotonate is used as a reactant for iminodipropionic acid as a leaving group for DNA polymerization by HIV-1 reverse transcriptase, orally bioavailable GPR40 agonist synthesis, preparation of mGlu4R agonists and synthesis of fluoroketone inhibitors of group VIA calcium-independent phospholipase A2.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text