2-Phenylbenzimidazole is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Phenylbenzimidazole is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
A.K.Mishra; S.K.Dogra. Effect of solvents and pH on the absorption and fluorescence spectra of 2-phenylbenzimidazole. Spectrochimica Acta Part A: Molecular Spectroscopy. 1983, 39,(7), 609-611.
Clarke Stevenson; R.J.H.Davies. Photosensitization of Gguanine-specific DNA damage by 2-phenylbenzimidazole and the sunscreen agent 2-phenylbenzimidazole-5-sulfonic acid. Chem. Res. Toxicol. 1999, 12,(1), 38-45.