2-Phenylbenzimidazole, 97%, Thermo Scientific Chemicals
2-Phenylbenzimidazole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Phenylbenzimidazole, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number A10996.22
also known as A10996-22
Price (USD)/ Each
212.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
212.00
Add to cart
2-Phenylbenzimidazole, 97%, Thermo Scientific Chemicals
Catalog numberA10996.22
Price (USD)/ Each
212.00
-
Add to cart
Chemical Identifiers
CAS716-79-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to brown
FormPowder
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%
2-Phenylbenzimidazole is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Phenylbenzimidazole is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. A.K.Mishra; S.K.Dogra. Effect of solvents and pH on the absorption and fluorescence spectra of 2-phenylbenzimidazole. Spectrochimica Acta Part A: Molecular Spectroscopy. 1983, 39,(7), 609-611.
  2. Clarke Stevenson; R.J.H.Davies. Photosensitization of Gguanine-specific DNA damage by 2-phenylbenzimidazole and the sunscreen agent 2-phenylbenzimidazole-5-sulfonic acid. Chem. Res. Toxicol. 1999, 12,(1), 38-45.