Thermo Scientific Chemicals

Benzylamine, 98+%, Thermo Scientific Chemicals

Catalog number: A10997.0I
5000 g, Each
Thermo Scientific Chemicals

Benzylamine, 98+%, Thermo Scientific Chemicals

Catalog number: A10997.0I
5000 g, Each
Quantity
Catalog number: A10997.0I
also known as A10997-0I
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-46-9
IUPAC Name
1-phenylmethanamine
Molecular Formula
C7H9N
InChI Key
WGQKYBSKWIADBV-UHFFFAOYSA-N
SMILES
NCC1=CC=CC=C1
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless
Assay (GC)
≥98.0%
Refractive Index
1.5415-1.5455 @ 20?C
Form
Liquid

Description

Benzylamine is a valuable intermediate for various applications and a building block for chemical synthesis used in pharmaceuticals and crop protection agents. It finds application in the coating industry. It is involved as a carrier electrolyte for separation of alkali, alkaline earth and ammonium cations in water samples by capillary electrophoresis with indirect UV detection. It is also used in synthesis of cross-linked porous copolymer supports based on N-(p-vinylbenzoyl)-2-methylalanine, styrene and divinylbenzene. In the textile industry, it is used in colored dyes. It is often used for medicinal purposes in topical creams and antifungal solutions as well as in vitamins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzylamine is a valuable intermediate for various applications and a building block for chemical synthesis used in pharmaceuticals and crop protection agents. It finds application in the coating industry. It is involved as a carrier electrolyte for separation of alkali, alkaline earth and ammonium cations in water samples by capillary electrophoresis with indirect UV detection. It is also used in synthesis of cross-linked porous copolymer supports based on N-(p-vinylbenzoyl)-2-methylalanine, styrene and divinylbenzene. In the textile industry, it is used in colored dyes. It is often used for medicinal purposes in topical creams and antifungal solutions as well as in vitamins.

Solubility
Miscible with water, ethanol, diethyl ether, acetone, benzene and chloroform.

Notes
Air sensitive. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text