The compound is a prolyl 4-hydroxylase inhibitor and can be used to protect the myocardium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The compound is a prolyl 4-hydroxylase inhibitor and can be used to protect the myocardium.
Solubility
Insoluble in water. Soluble in ethanol.
Notes
Keep container tightly closed in a cool, dry and well-ventilated place. Store away from oxidizing agents, Strong bases.
RUO – Research Use Only
Bianca L. Garner.; Jean E.L. Arceneaux.; Rowe Byers, B. Temperature Control of a 3,4-Dihydroxybenzoate (Protocatechuate)-Based Siderophore in Bacillus anthracis. Current Microbiology. 2000, 49, (2), 89-94.
Osamu Saitoh.; Muthu Periasamy.; Mikio Kan.; Ryoichi Matsuda. Cis-4-hydroxy-l-proline and ethyl-3,4-dihydroxybenzoate prevent myogenesis of C2C12 muscle cells and block myoD1 and myogenin expression. Experimental Cell Research. 1992, 200, (1), 70-76.