Thermo Scientific Chemicals

Ethyl N-hydroxyacetimidate, 97%, Thermo Scientific Chemicals

Catalog number: A11006.06
5 g, Each
Thermo Scientific Chemicals

Ethyl N-hydroxyacetimidate, 97%, Thermo Scientific Chemicals

Catalog number: A11006.06
5 g, Each
Quantity
Catalog number: A11006.06
also known as A11006-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10576-12-2
IUPAC Name
(E)-(ethyl N-hydroxyethanimidate)
Molecular Formula
C4H9NO2
InChI Key
QWKAVVNRCKPKNM-SNAWJCMRSA-N
SMILES
CCO\C(C)=N\O
Appearance (Color)
Colorless to white to pale yellow
Form
Fused solid or clear liquid as melt
Assay (GC)
≥96.0%
Refractive Index
1.4315-1.4365 @ 20?C

Description

Ethyl N-hydroxyacetimidate was used in the synthesis of O-acyl-and O-nitrophenylhydroxylamines, hydroxamic acid ethoxycarbonylhydrazides and ethyl O-(2,4-dinitrophenyl)acetohydroxamate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl N-hydroxyacetimidate was used in the synthesis of O-acyl-and O-nitrophenylhydroxylamines, hydroxamic acid ethoxycarbonylhydrazides and ethyl O-(2,4-dinitrophenyl)acetohydroxamate.

Solubility
Decompose in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text