Thermo Scientific Chemicals

Benzaldehyde dimethyl acetal, 99%, Thermo Scientific Chemicals

Catalog number: A11009.0E
2500 g, Each
Thermo Scientific Chemicals

Benzaldehyde dimethyl acetal, 99%, Thermo Scientific Chemicals

Catalog number: A11009.0E
2500 g, Each
Quantity
Catalog number: A11009.0E
also known as A11009-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1125-88-8
IUPAC Name
(dimethoxymethyl)benzene
Molecular Formula
C9H12O2
InChI Key
HEVMDQBCAHEHDY-UHFFFAOYSA-N
SMILES
COC(OC)C1=CC=CC=C1
Refractive Index
1.4910-1.4945 @ 20°C
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms

Description

Used in vegetable flavors such as parsley and carrots. For almond, nut, fruit, violet and other nutty and floral fragrances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in vegetable flavors such as parsley and carrots. For almond, nut, fruit, violet and other nutty and floral fragrances.

Solubility
Insoluble in water. Soluble in alcohol.

Notes
Keep container tightly closed in a cool, dry and well-ventilated place. Store away from oxidizing agents and acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text