Thermo Scientific Chemicals

Tetra-n-butylammonium trifluoromethanesulfonate, 99%, Thermo Scientific Chemicals

Catalog number: A11013.14
25 g, Each
Thermo Scientific Chemicals

Tetra-n-butylammonium trifluoromethanesulfonate, 99%, Thermo Scientific Chemicals

Catalog number: A11013.14
25 g, Each
Quantity
Catalog number: A11013.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
35895-70-6
Appearance (Color)
White to pale cream
Form
Crystalline powder
Purity (DSC)
≥98.5%
Melting Point (DSC)
111-113?C

Description

Used as a catalyst for the reactions of condensation of alcohols and carboxylic acids, reaction of aromatic compounds with sulfonyl chlorides, cracking of alkanes, alkylation of alkenes, isomerisation of alkanes and trans-alkylation of aromatics, trans-bromination and other Friedel-Crafts reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a catalyst for the reactions of condensation of alcohols and carboxylic acids, reaction of aromatic compounds with sulfonyl chlorides, cracking of alkanes, alkylation of alkenes, isomerisation of alkanes and trans-alkylation of aromatics, trans-bromination and other Friedel-Crafts reactions.

Solubility
Miscible in watetr. Soluble in methanol(0.1 g/mL).

Notes
Hygroscopic. Keep container tightly closed in a cool, dry and well-ventilated place. Store away from oxidizing agents, air and water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text