Used as a catalyst for the reactions of condensation of alcohols and carboxylic acids, reaction of aromatic compounds with sulfonyl chlorides, cracking of alkanes, alkylation of alkenes, isomerisation of alkanes and trans-alkylation of aromatics, trans-bromination and other Friedel-Crafts reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as a catalyst for the reactions of condensation of alcohols and carboxylic acids, reaction of aromatic compounds with sulfonyl chlorides, cracking of alkanes, alkylation of alkenes, isomerisation of alkanes and trans-alkylation of aromatics, trans-bromination and other Friedel-Crafts reactions.
Solubility
Miscible in watetr. Soluble in methanol(0.1 g/mL).
Notes
Hygroscopic. Keep container tightly closed in a cool, dry and well-ventilated place. Store away from oxidizing agents, air and water.
RUO – Research Use Only
Kenneth G. Furton.; Colin F. Poole. Variation in the gas chromatographic stationary phase propertiesof tetra-n-butylammonium salts as a function of the anion type. Journal of Chromatography A. 1985, 349, (2), 235-247.
Piersona, J.F.; Wiederkehra, D.; Billard, A. Correlation of solute retention in gas chromatography with properties of the anion for tetra-n-butylammonium salts. J. Org. Chem. 2005, 478, (1-2), 196-205.
Supporting electrolyte for organic electrochemistry in aprotic solvents, having good solubility and a high degree of chemical inertness: J. Org. Chem., 37, 3968 (1972).