2-Fluorobenzoic acid may be employed in the preparation of zaragozic acid A analogs. It is also used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Fluorobenzoic acid may be employed in the preparation of zaragozic acid A analogs. It is also used as an organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
T.S.Chen; B.Petuch; J.MacConnell; R.White; G.Dezeny; B.Arison; J.D.Bergstrom; L.Colwell; L.Huang; R.L.Monaghan. The preparation of zaragozic acid A analogues by directed biosynthesis. Journal of Antibiotics. 1994, 47,(11), 1290-1294.
Peter Goldman; G.W.A.Milne; Maria T.Pignataro. Fluorine containing metabolites formed from 2-fluorobenzoic acid by Pseudomonas species. Archives of Biochemistry and Biophysics. 1967, 118,(1), 178-184.