Thermo Scientific Chemicals

2-Fluorobenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A11015.36
500 g, Each
Thermo Scientific Chemicals

2-Fluorobenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A11015.36
500 g, Each
Quantity
Catalog number: A11015.36
also known as A11015-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
445-29-4
IUPAC Name
2-fluorobenzoic acid
Molecular Formula
C7H5FO2
InChI Key
NSTREUWFTAOOKS-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CC=C1F
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0% (UK Sourced Material)
Assay (Silylated GC)
≥98.0% (UK Sourced Material)
Assay (HPLC)
≥98.0% (UK Sourced Material)

Description

2-Fluorobenzoic acid may be employed in the preparation of zaragozic acid A analogs. It is also used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluorobenzoic acid may be employed in the preparation of zaragozic acid A analogs. It is also used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text