4-Aminobutyric acid, 97%, Thermo Scientific Chemicals
4-Aminobutyric acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Aminobutyric acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2500 g
Catalog number A11016.36
also known as A11016-36
Price (USD)/ Each
180.65
Online exclusive
200.00 
Save 19.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
180.65
Online exclusive
200.00 
Save 19.35 (10%)
Add to cart
4-Aminobutyric acid, 97%, Thermo Scientific Chemicals
Catalog numberA11016.36
Price (USD)/ Each
180.65
Online exclusive
200.00 
Save 19.35 (10%)
-
Add to cart
Chemical Identifiers
CAS56-12-2
IUPAC Name4-aminobutanoic acid
Molecular FormulaC4H9NO2
InChI KeyBTCSSZJGUNDROE-UHFFFAOYSA-N
SMILESNCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%
Water Content (Karl Fischer Titration)≤1.5%
Appearance (Color)White
FormPowder
In the production of polymers, food additives, and pharmaceuticals. Acrylic acid and methacrylic acids are well known precursors of polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In the production of polymers, food additives, and pharmaceuticals. Acrylic acid and methacrylic acids are well known precursors of polymers.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. A. W. Bown,; B. J. Shelp.The Metabolism and Functions of [gamma]-Aminobutyric Acid. Plant Physiol. 1997, 115 (1),1-5
  2. Sadaji Yokoyama,; Jun-ichi Hiramatsu1,; Kiyoshi Hayakawa,; Michael D McLeana.Production of γ-aminobutyric acid from alcohol distillery lees by Lactobacillus brevis IFO-12005. Journal of Bioscience and Bioengineering. 2002, 93(1),95-97.