Thermo Scientific Chemicals

Isobutylamine, 99%, Thermo Scientific Chemicals

Catalog number: A11017.AU
1000 mL, Each
Thermo Scientific Chemicals

Isobutylamine, 99%, Thermo Scientific Chemicals

Catalog number: A11017.AU
1000 mL, Each
Quantity
Catalog number: A11017.AU
also known as A11017-AU
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
78-81-9
IUPAC Name
2-methylpropan-1-amine
Molecular Formula
C4H11N
InChI Key
KDSNLYIMUZNERS-UHFFFAOYSA-N
SMILES
CC(C)CN
Form
Liquid
Assay (GC)
>98.5%
Water Content (Karl Fischer Titration)
<0.5%
Refractive Index
1.3955-1.3995 @ 20?C
Appearance (Color)
Clear, colorless

Description

Isobutylamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isobutylamine is used as pharmaceutical intermediate.

Solubility
Soluble in water (1000 mg/ml at 25°C), ether, alcohol, acetone and benzene.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, acids, acid chlorides and acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text